Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124421

Catalog No. PIPA124421


Add to cart

Description

Description

PA1-24421 detects Claudin 9 in Human samples. PA1-24421 has been successfully used in Western Blot procedures. Dilute to 2.0 µg/mL in 5% skim milk/PBS buffer. PA1-24421 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human CLDN9. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

CLDN9 is a member of claudin family. It is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C.
Specifications

Specifications

Claudin 9
Polyclonal
Unconjugated
CLDN9
Liquid
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
9080
Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
O95484
CLDN9
Synthetic peptide region of human CLDN9.
Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit