Learn More
anti-CLCN7, Polyclonal, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP130854
Catalog No. NBP130854
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
CLCN7
LYOPH
Western Blot 1:300-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:300 - 1:2000, Immunohistochemistry-Frozen 1:300 - 1:2000
Antisera
CLCN7
A synthetic peptide from the cytoplasmic domain of human Chloride channel protein 7 (CLCN7) conjugated to an immunogenic carrier protein was used as the immunogen.
Unpurified
RUO
Primary
1186
Reconstitute with 0.1 ml sterilized water to desired concentration.
Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Whole antisera with No Preservative
chloride channel 7, Chloride channel protein 7, CLC-7, CLC7FLJ39644, FLJ26686, H(+)/Cl(-) exchange transporter 7, OPTA2FLJ46423, OPTB4
Rabbit
IgG
0.1 ml
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Appears to be specific for CLCN7.
Human, Mouse, Rat