Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Claudin 12 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA525768
Description
This antibody is predicted to react with bovine and mouse based on sequence homology.
The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands.Specifications
Claudin 12 | |
Polyclonal | |
Unconjugated | |
P56749 | |
CLDN12 | |
KLH conjugated synthetic peptide between 19-46 amino acids from the N-terminal region of human CLDN12 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
0.43 mg/mL | |
CLDN12 | |
claudin-12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9069 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction