Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 15 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526830

Catalog No. PIPA526830


Add to cart

Description

Description

Plays a major role in tight junction specific obliteration of the intercellular space, through calcium independent cell adhesion activity.
Specifications

Specifications

Claudin 15
Polyclonal
PBS with 0.09% sodium azide
P56746
claudin-15
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
24146
Flow Cytometry, Western Blot
Unconjugated
CLDN15
Liquid
CLDN15
KLH conjugated synthetic peptide between 128-157 amino acids from the central region of human CLDN15
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit