Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 18 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526237

Catalog No. PIPA526237

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CLDN18 belongs to the large claudin family of proteins, which form tight junction strands in epithelial cells.
Specifications

Specifications

Claudin 18
Polyclonal
PBS with 0.09% sodium azide
P56856
SFTA5, SFTPJ, claudin-18, surfactant associated 5, surfactant associated protein J, surfactant, pulmonary associated protein J
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
51208
Flow Cytometry, Western Blot
Unconjugated
CLDN18
Liquid
CLDN18
KLH conjugated synthetic peptide between 231-259 amino acids from the C-terminal region of human CLDN18
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit