Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 22 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523738

Catalog No. PIPA523738


Add to cart

Description

Description

This gene encodes a member of the claudin family. Claudins are integral membrane proteins and components of tight junction strands. Tight junction strands serve as a physical barrier to prevent solutes and water from passing freely through the paracellular space between epithelial or endothelial cell sheets, and also play critical roles in maintaining cell polarity and signal transductions. This gene is intronless and overlaps the 3 ft. UTR of the WWC2 gene (GeneID: 80014) on the opposite strand.
Specifications

Specifications

Claudin 22
Polyclonal
Unconjugated
CLDN22
Liquid
CLDN22
KLH conjugated synthetic peptide between 90-117 amino acids from the central region of human CLDN22
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Western Blot
0.25 mg/mL
PBS with 0.09% sodium azide
Q8N7P3, Q9D7U6
CLDN21, claudin-22
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
53842, 75677
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit