Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 23 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526831

Catalog No. PIPA526831

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CLDN23 is a four-transmembrane protein with WWCC motif, defined by W-X(17-22)-W-X(2)-C-X(8-10)-C. It is a candidate tumor suppressor gene implicated in intestinal-type gastric cancer.
Specifications

Specifications

Claudin 23
Polyclonal
PBS with 0.09% sodium azide
Q96B33, Q9D7D7
CLDNL, hCG1646163, 2310014B08Rik, claudin-23
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
137075, 71908
Immunohistochemistry, Western Blot
Unconjugated
CLDN23
Liquid
CLDN23
KLH conjugated synthetic peptide between 228-257 amino acids from the C-terminal region of human CLDN23
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit