Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 4 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526621

Catalog No. PIPA526621


Add to cart

Description

Description

CLDN4 is an integral membrane protein, which belongs to the claudin family. The protein is a component of tight junction strands and may play a role in internal organ development and function during pre- and postnatal life.
Specifications

Specifications

Claudin 4
Polyclonal
PBS with 0.09% sodium azide
O14493, O35054
CPE-R, CPER, CPETR, CPETR1, WBSCR8, hCPE-R, CPE-receptor, Clostridium perfringens enterotoxin receptor 1, Williams-Beuren syndrome chromosomal region 8 protein, claudin-4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
12740, 1364
Western Blot
Unconjugated
CLDN4
Liquid
CLDN4
KLH conjugated synthetic peptide between 171-197 amino acids from the C-terminal region of human CLDN4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit