Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 8 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526231

Catalog No. PIPA526231

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CLDN8 is components of epithelial cell tight junctions. Tight junctions regulate movement of solutes and ions through the paracellular space and prevent mixing of proteins and lipids in the outer leaflet of the apical and basolateral plasma membrane domains
Specifications

Specifications

Claudin 8
Polyclonal
PBS with 0.09% sodium azide
P56748
claudin-8
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9073
Flow Cytometry, Western Blot
Unconjugated
CLDN8
Liquid
CLDN8
KLH conjugated synthetic peptide between 140-166 amino acids from the central region of human CLDN8
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit