Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLDND1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526733

Catalog No. PIPA526733


Add to cart

Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

CLDND1 is widely distributed in the adult CNS with highest expression in the corpus callosum, caudate nucleus, cerebral cortex, medulla, putamen, spinal cord, substantia nigra and subthalamic nucleus. Weak expression has been detected in the adult heart. There are 2 named isoforms produced by alternative splicing.
Specifications

Specifications

CLDND1
Polyclonal
PBS with 0.09% sodium azide
Q9NY35, Q9CQX5
C3orf4, GENX-3745, claudin domain containing 1 protein, claudin domain-containing protein 1, membrane protein GENX-3745
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
224250, 56650
Western Blot
Unconjugated
CLDND1
Liquid
CLDND1
KLH conjugated synthetic peptide between 43-72 amino acids from the central region of human CLDND1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit