Learn More
CSTF2 Mouse anti-Human, Clone: 4E3-3C9, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA518668
Catalog No. PIMA518668
Add to cart
Description
Description
Peptide Sequence: MAGLTVRDPA VDRSLRSVFV GNIPYEATEG QLKDIFSEVG PVVSFRLVYD RETGKPKGYG FCEYQDQETA LNAMRNLNGR EFSGRALRVD NAASEKNKEE LKSLGTGAPV IESPYGETIS PEDAPESISK AVASLPPEQM FELMKQMKLC VQNSPQEARN MLLQNPQLAY ALLQAQVVMR IVDPEIALKI LHRQTNIPTL IAGNPQPVHG AGPGSGSNVS MNQQNPQAPQ AQSLGGMHVN GAPPLMQASM QGGVPAPGQM PAAVTGPGPG SLAPGGGMQA QVGMPGSGPV SMERGQVPMQ DPRAAMQRGS LPANVPTPRG LLGDAPNDPR GGTLLSVTGE VEPRGYLGPP HQGPPMHHVP GHESRGPPPH ELRGGPLPEP RPLMAEPRGP MLDQRGPPLD GRGGRDPRGI DARGMEARAM EARGLDARGL EARAMEARAM EARAMEVRGM EARGMDTRGP VPGPRGPIPS GMQGPSPINM GAVVPQGSRQ VPVMQGTGMQ GASIQGGSQP GGFSPGQNQV TPQDHEKAAL IMQVLQLTAD QIAMLPPEQR QSILILKEQI QKSTGAP
This gene encodes a nuclear protein with an RRM (RNA recognition motif) domain. The protein is a member of the cleavage stimulation factor (CSTF) complex that is involved in the 3 ft. end cleavage and polyadenylation of pre-mRNAs. Specifically, this protein binds GU-rich elements within the 3 ft.-untranslated region of mRNAs.
Specifications
Specifications
CSTF2
Monoclonal
1.0 mg/mL
PBS with no preservative; pH 7.4
P33240
CstF-64, CF-1 64 kDa subunit, CSTF 64 kDa subunit, betaCstF-64 variant 2, cleavage stimulation factor 64 kDa subunit, cleavage stimulation factor subunit 2
Mouse
IgG1, kappa
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1478
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
4E3-3C9
Unconjugated
CSTF2
Liquid
CSTF2
CSTF2 (AAH17712, 1 a.a. ∽ 577 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Affinity chromatography
RUO
Antibody
Monoclonal
Human