Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD301 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524058

Catalog No. PIPA524058


Add to cart

Description

Description

CLEC10A encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may function as a cell surface antigen. Two transcript variants encoding distinct isoforms have been identified for this gene.
Specifications

Specifications

CD301
Polyclonal
PBS with 0.09% sodium azide
Q8IUN9
CD301, CLECSF13, CLECSF14, HML, HML2, MGL
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10462
Flow Cytometry, Western Blot
Unconjugated
CLEC10A
Liquid
CLEC10A
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human CLEC10A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit