Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC2A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524536

Catalog No. PIPA524536

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CLEC2A belongs to the CLEC2 family of activation-induced, natural killer gene complex-encoded C-type lectin-like receptors.
Specifications

Specifications

CLEC2A
Polyclonal
PBS with 0.09% sodium azide
Q6UVW9
KACL, PILAR, UNQ5792, C-type lectin domain family 2 member A, INPE5792, keratinocyte-associated C-type lectin, proliferation-induced lymphocyte-associated receptor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
387836
Western Blot
Unconjugated
CLEC2A
Liquid
CLEC2A
KLH conjugated synthetic peptide between 79-107 amino acids from the central region of human CLEC2A
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit