Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC2L Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524394

Catalog No. PIPA524394

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with rat based on sequence homology.

Specifications

Specifications

CLEC2L
Polyclonal
PBS with 0.09% sodium azide
P0C7M8, P0C7M9
C-type lectin domain family 2 member L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
154790, 665180
Western Blot
Unconjugated
CLEC2L
Liquid
CLEC2L
KLH conjugated synthetic peptide between 26-54 amino acids from the N-terminal region of human CLEC2L
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit