Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC4D Mouse anti-Human, Clone: 413512, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA524152

Catalog No. PIMA524152


Add to cart

Description

Description

In direct ELISAs and Western blots, no cross-reactivity with recombit human (rh) CLECSF9, rhCLECSF13, rhOCIL, or rhOCILrp2 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.

Specifications

Specifications

CLEC4D
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
Q8WXI8
CLEC6, CLECSF8, Mpcl, CLEC6, CLEC-6, CLECSF8Mpcl, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 8, C-type lectin domain family 4 member D, C-type lectin domain family 4, member D, C-type lectin receptor, C-type lectin superfamily member 8, C-type lectin-like receptor 6, macrophage C-type lectin, MCL, MGC40078, MPCL
Mouse
IgG2B
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
338339
Flow Cytometry, Western Blot
413512
Unconjugated
CLEC4D
Lyophilized
CLEC4D
Mouse myeloma cell line NS0-derived recombit human CLEC4D/CLECSF8 Gly52-Asn215
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit