Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLEC5A Monoclonal Antibody (283834), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA523909
Description
This antibody does not cross-react with recombinant mouse MDL-1. Reconstitute at 0.5 mg/mL in sterile PBS.
CLEC5A Monoclonal antibody specifically detects CLEC5A in Human samples. It is validated for Flow Cytometry, Western Blot.Specifications
CLEC5A | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9NY25 | |
CLEC5A | |
Mouse myeloma cell line NS0-derived recombinant human MDL-1/CLEC5A Tyr26-Lys188 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Western Blot | |
283834 | |
Unconjugated | |
CLEC5A | |
CLEC5A; Clecsf5; C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamily member 5; C-type lectin domain family 5 member A; C-type lectin domain family 5, member a; C-type lectin superfamily member 5; Ly100; Mdl1; MDL-1; myeloid DAP12-associating lectin 1; myeloid DAP12-associating lectin-1; type II transmembrane protein MDL-1 | |
Mouse | |
Protein A/G | |
RUO | |
23601 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction