Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLEC6 Mouse anti-Human, PE, Clone: 9B9 (also known as AB7_10.9B9), BD

Mouse Monoclonal Antibody

Manufacturer:  BD Biosciences 564413

Catalog No. BDB564413


Add to cart

Description

Description

The 9B9 monoclonal antibody specifically binds to CLEC6 (C-type lectin-like receptor 6) which is also known as CLEC4D, CLECSF8, and MCL (macrophage C-type lectin). CLEC6 is a 30 kDa type II transmembrane glycoprotein that belongs to the C-type Lectin Receptor family of molecules. It is primarily expressed on peripheral blood neutrophils and monocytes and weakly on several subsets of blood dendritic cells. Expression of this receptor is lost upon in vitro differentiation of monocytes into dendritic cells or macrophages. CLEC6 reportedly serves as an endocytic receptor that may function in antigen clearance and presentation to T lymphocytes. CLEC6 may also play roles in cellular adhesion, signaling and glycoprotein turnover. CLEC6 expression can be upregulated by cells in response to cytokines including IFN-γ, TNF, IL-6, and IL-10.

Specifications

Specifications

CLEC6
9B9 (also known as AB7_10.9B9)
CD368; CLEC4D; CLECSF8; MCL; CLEC6
Affinity Purified
Human CLEC6 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Flow Cytometry
PE
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
IgG2b, κ
100 Tests
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit