Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clec9A Rat anti-Human, Alexa Fluor 647, Clone: 3A4/Clec9A, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 564267

Catalog No. BDB564267


Add to cart

Description

Description

The 3A4/Clec9A monoclonal antibody specifically binds to human Clec9A (C-type lectin domain family member 9A). Clec9A is a type II transmembrane glycoprotein with a single extracellular C-type lectin domain. Clec9A is also known as CD370 and DNGR1 (Dendritic cell natural killer lectin group receptor 1). Clec9A is selectively expressed on CD141+ myeloid dendritic cells. Clec9A reportedly functions as an endocytic receptor for necrotic cells. It can mediate the cross-presentation of dead-cell associated antigens by dendritic cells in a Syk-dependent manner. The 3A4 antibody reportedly crossreacts with peripheral blood dendritic cells form Pigtail macaques ( Macaca nemestrina ).

Specifications

Specifications

Clec9A
3A4/Clec9A
Alexa Fluor 647
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
RUO
Primary
Human
Flow Cytometry
0.2mg/mL
CD370; C-type lectin domain family 9 member A; CLEC9A; DNGR1
Affinity Purified
Human Clec9A Peptide
25μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit