Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-CLEC9a, APC, Clone: 683409, R&D Systems™
Mouse Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems FAB6049A
This item is not returnable.
View return policy
Description
CLEC9a Monoclonal specifically detects CLEC9a in Human samples. It is validated for Flow Cytometry.Specifications
CLEC9a | |
Monoclonal | |
APC | |
Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
CD370, C-type lectin domain family 9 member A, C-type lectin domain family 9, member A, DNGR1, HEEE9341, UNQ9341 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
283420 | |
Human | |
Purified |
Flow Cytometry | |
683409 | |
Flow Cytometry 10 uL/10^6 cells | |
Q6UXN8 | |
CLEC9A | |
Mouse myeloma cell line NS0-derived recombinant human CLEC9a Lys57-Val241 Accession # Q6UXN8 | |
100 Tests | |
Primary | |
Detects human CLEC9a in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human (rh) CLEC1, 2, 2A, 3B, 10A, 12B, 14A, rhCD302/CLEC13a, rhMICL, or recombinant mouse CLEC9a is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title