Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLIC6 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523976
Description
CLIC6 is a member of the chloride intracellular channel family of proteins. The gene is part of a large triplicated region found on chromosomes 1, 6, and 21. An alternatively spliced transcript variant has been described, but its biological validity has not been determined.Specifications
CLIC6 | |
Polyclonal | |
CLIC6 | |
CLIC1L, chloride channel form A, chloride intracellular channel protein 6, parchorin | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54102 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
Unconjugated | |
Q96NY7 | |
CLIC6 | |
KLH conjugated synthetic peptide between 539-565 amino acids from the central region of human CLIC6 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction