Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLIF, Polyclonal, Invitrogen™

Catalog No. PIPA127938
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
PIPA127938 50 μg
1 options

Catalog No. PIPA127938

Supplier: Thermo Scientific PA127938

Chicken Polyclonal Antibody

PA1-27938 detects CLIF from human samples.PA1-27938 may be successfully used in Western blot applications. By Western blot, this antibody has only been tested on fusion protein.The PA1-27772 immunogen is a synthetic peptide corresponding to human Cycle-like factor CLIF.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLIF
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene ARNTL2
Gene Accession No. Q8WYA1
Gene Alias BMAL2, CLIF, MGC149671, MGC149672, MOP9, PASD9, bHLHe6
Gene Symbols ARNTL2
Host Species Chicken
Immunogen Synthetic peptide corresponding to human Cycle-like factor CLIF.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56938
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.