Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CLIP3 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA524559
Description
This antibody is predicted to react with mouse based on sequence homology.
Functions as a cytoplasmic linker protein. Involved in TGN-endosome dynamics.
Specifications
CLIP3 | |
Polyclonal | |
CLIP3 | |
CLIPR-59, CLIPR59, RSNL1, CAP-Gly domain-containing linker protein 3, CLIP-170-related 59 kDa protein, cytoplasmic linker protein 170-related 59 kDa protein, restin-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
25999 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
Unconjugated | |
Q96DZ5 | |
CLIP3 | |
KLH conjugated synthetic peptide between 368-396 amino acids from the C-terminal region of human CLIP3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction