Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD300f Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526947

Catalog No. PIPA526947

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

CLM1 is an inhibitory receptor of the Ig superfamily expressed on myeloid cells. It mediates negative regulatory signals by recruiting SHP1 (PTPN6; MIM 176883) or SHIP.
Specifications

Specifications

CD300f
Polyclonal
PBS with 0.09% sodium azide
Q8TDQ1
CD300f, CLM-1, CLM1, IREM-1, IREM1, IgSF13, NKIR, CD300 antigen like family member F, CD300 antigen-like family member F, CD300 molecule like family member f, CMRF35-like molecule 1, NK inhibitory receptor, immune receptor expressed on myeloid cells 1, immunoglobin superfamily member 13, immunoglobulin superfam
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
146722
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CD300LF
Liquid
CD300LF
KLH conjugated synthetic peptide between 56-84 amino acids from the N-terminal region of human CLM1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit