Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clostridia Rabbit anti-Bacteria, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA17210

Catalog No. PA17210


Add to cart

Description

Description

PA1-7210 detects clostridium. PA1-7210 has been successfully used in immunofluorescence procedures. The PA1-7210 immunogen is extract of Cl. perfingens, Cl. sordellii, Cl. novyi, Cl. septicum, Cl. chauvoei.

Specifications

Specifications

Clostridia
Polyclonal
Unconjugated
Liquid
Rabbit
IgG
1 mL
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Primary
Bacteria
Immunofluorescence
4-5 mg/mL
PBS with 0.1% sodium azide
Clostridium
Extract of Cl. perfingens, Cl. sordellii, Cl. novyi, Cl. septicum, Cl. chauvoei.
Purified
RUO
Antibody
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit