Learn More
Clostridium tetani Tetanus Toxin Mouse anti-Bacteria, Clone: 6E7, Invitrogen
Mouse Monoclonal Antibody
Manufacturer: Invitrogen HYB2781702
Description
HYB 278-17-02 detects tetanus toxin and tetanus toxoid. HYB 278-17-02 has been successfully used in ELISA procedures. C. tetani toxoid injected in mice is neutralized by administration of HYB 278-17-02. The HYB 278-17-02 immunogen is Tetanus toxoid, formaldehyde inactivated Tetanus toxin. Epitope specificity of HYB 278-17-02 differs from that of HYB 278-14-02 and HYB 278-15-02. NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
Tetanus toxin is a neurotoxin produced by the vegetative spore of Clostridium tetani. The toxin causes tetanus, a disease that is characterised by prolonged contraction of skeletal muscle. World-wide, approximately 1 million cases of tetanus occurs annually, resulting in approximately 300,000 to 500,000 deaths, mostly in developing countries. The tetanus toxin has a molecular weight of approximately 150kDa and consists of a heavy chain (B-chain, 100kDa) and a light chain (A-chain, 50kDa) that are connected by a disulfide bond. The toxin acts by inhibiting neurotransmitter release. It binds to peripheral neuronal synapses, is internalized and moves by retrograde transport up the axon into the spinal cord where it can move between postsynaptic and presynaptic neurons. It inhibits the release of the neurotransmitters GABA and glycine by synaptobrevin-2. This results in overactivity in the muscle. The binding of the toxin to neurons is irreversible.Specifications
Clostridium tetani Tetanus Toxin | |
Monoclonal | |
1.06 mg/mL | |
PBS with 15mM sodium azide; pH 7.4 | |
C. tetani; Tetanus Toxin | |
Tetanus toxoid, formaldehyde inactivated Tetanus toxin | |
Protein A | |
RUO | |
Antibody | |
Monoclonal |
ELISA | |
6E7 | |
Unconjugated | |
Liquid | |
Mouse | |
IgG1, kappa | |
200 μL | |
4° C, store in dark | |
Primary | |
Bacteria |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.