Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLPTM1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526627

Catalog No. PIPA526627

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with bovine and zebrafish based on sequence homology.

CLPTM1 may play a role in T-cell development.
Specifications

Specifications

CLPTM1
Polyclonal
PBS with 0.09% sodium azide
O96005, Q8VBZ3
cleft lip and palate transmembrane protein 1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1209, 56457
Western Blot
Unconjugated
CLPTM1
Liquid
CLPTM1
KLH conjugated synthetic peptide between 198-227 amino acids from the central region of human CLPTM1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit