Learn More
CLPX Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA524256
Catalog No. PIPA524256
Add to cart
Description
Description
This antibody is predicted to react with rat based on sequence homology.
ATP-dependent specificity component of the Clp protease. It directs the protease to specific substrates. Can perform chaperone functions in the absence of clpP.
Specifications
Specifications
CLPX
Polyclonal
Unconjugated
CLPX
Liquid
CLPX
KLH conjugated synthetic peptide between 449-477 amino acids from the C-terminal region of human CLPX
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.4 mg/mL
PBS with 0.09% sodium azide
O76031, Q9JHS4
ATP-dependent Clp protease ATP-binding subunit clpX-like, mitochondrial, ClpX caseinolytic protease X homolog, energy-dependent regulator of proteolysis
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10845, 270166