Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Light Chain A Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526026

Catalog No. PIPA526026

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Clathrin is a large, soluble protein composed of heavy and light chains. It functions as the main structural component of the lattice-type cytoplasmic face of coated pits and vesicles which entrap specific macromolecules during receptor-mediated endocytosis. CLTA is one of two clathrin light chain proteins which are believed to function as regulatory elements.
Specifications

Specifications

Clathrin Light Chain A
Polyclonal
PBS with 0.09% sodium azide
P09496
LCA, clathrin light chain A, clathrin, light polypeptide (Lca)
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1211
Flow Cytometry, Immunohistochemistry, Western Blot
Unconjugated
CLTA
Liquid
CLTA
KLH conjugated synthetic peptide between 99-127 amino acids from the central region of human CLTA
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit