Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Clathrin Light Chain B, Polyclonal, Invitrogen™

Chicken Polyclonal Antibody

Manufacturer:  Invitrogen PA127770

Catalog No. PIPA127770

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Clathrin is the main structural component of the lattice covering the cytoplasmic face of the coated pits and coated vesicles in which specific macromolecules are entrapped

PA1-27770 detects CLTB from human samples.PA1-27770 may be successfully used in ELISA and Western blot applications.The PA1-27695 immunogen is a synthetic peptide corresponding to human Clathrin light chain B.
Specifications

Specifications

CLTB
Polyclonal
PBS with no preservative
P09497
LCB
Chicken
IgY
50μg
Store at -20°C
Primary
1212
ELISA, Western Blot
Unconjugated
CLTB
Purified
CLTB
Synthetic peptide corresponding to human Clathrin light chain B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit