Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clusterin α chain Mouse anti-Human, Clone: 41D, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 05354
Description
Anti-Clusterin α chain (human) Antibody, clone 41D is an antibody against Clusterin α chain (human) for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.Specifications
Clusterin α chain | |
Monoclonal | |
Unconjugated | |
NM_203339 | |
Mouse | |
Protein G purfied | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
41D | |
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, with 0.05% sodium azide. Frozen at −20°C. | |
CLU, SP-40, MGC24903, NA1/NA2, SGP-2, AAG4, KUB1, APOJ, CLI, TRPM-2, TRPM2, Apo-J, SGP2 | |
Native clusterin purified from human serum. Clone 41D. | |
100 μg | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only