Learn More
anti-CMAS, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP232324
Catalog No. NBP232324
Add to cart
Description
Description
CMAS Polyclonal antibody specifically detects CMAS in Human samples. It is validated for Western Blot.
Specifications
Specifications
CMAS
0.42 mg/mL
Western Blot 1:1000
Q8NFW8
CMP-N-acetylneuraminic acid synthase, CMP-N-acetylneuraminic acid synthetase, CMP-Neu5Ac synthetase, CMP-NeuNAc synthase, CMP-NeuNAc synthetase, cytidine 5'-monophosphate N-acetylneuraminic acid synthetase, cytidine monophosphate N-acetylneuraminic acid synthetase, EC 2.7.7.43, N-acylneuraminate cytidylyltransferase
Rabbit
IgG
Protein A purified
RUO
Primary
55907
Western Blot
Unconjugated
PBS with 0.09% Sodium Azide
Affinity Purified
CMAS
This CMAS antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 300-327 amino acids from the C-terminal region of human CMAS.
48.379 kDa
400 ul
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human