Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-CMAS, Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP132418

Catalog No. NBP132418

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

CMAS
1.0mg/mL
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Affinity Purified
CMAS
Recombinant protein encompassing a sequence within the center region of human CMAS. The exact sequence is proprietary.
Immunogen affinity purified
RUO
Primary
55907
Human
Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
CMP-N-acetylneuraminic acid synthase, CMP-N-acetylneuraminic acid synthetase, CMP-Neu5Ac synthetase, CMP-NeuNAc synthase, CMP-NeuNAc synthetase, cytidine 5'-monophosphate N-acetylneuraminic acid synthetase, cytidine monophosphate N-acetylneuraminic acid synthetase, EC 2.7.7.43, N-acylneuraminate cytidylyltransferase
Rabbit
IgG
100 ul
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Polyclonal
Tested in human. Predicted cross-reactivity based on sequence identity: Rhesus Monkey (100%). Not yet tested in any other species.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only