Learn More
CMAS Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA521654
Catalog No. PIPA521654
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA5-21656 targets BRE in ICC, IF, IP, and WB applications and shows reactivity with Human and mouse samples. The PA5-21656 immunogen is recombit protein fragment contain a sequence corresponding to a region within amino acids 9 and 328 (Q9NXR7) of BRE.
Specifications
Specifications
CMAS
Polyclonal
Unconjugated
CMAS
Liquid
CMAS
Recombit fragment corresponding to a region within amino acids 69 and 414 of CMAS (Uniprot IDNo.Q8NFW8)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Q8NFW8
CMP-N-acetylneuraminic acid synthase, CMP-N-acetylneuraminic acid synthetase, CMP-Neu5Ac synthetase, CMP-NeuNAc synthase, CMP-NeuNAc synthetase, N-acylneuraminate cytidylyltransferase, cytidine 5′-monophosphate N-acetylneuraminic acid synthetase
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55907