Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNNM4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA525334

Catalog No. PIPA525334


Add to cart

Description

Description

This antibody is predicted to react with bovine, chicken and rat based on sequence homology.

This gene encodes a member of the ancient conserved domain containing protein family. Members of this protein family contain a cyclin box motif and have structural similarity to the cyclins. The encoded protein may play a role in metal ion transport. Mutations in this gene are associated with Jalili syndrome which consists of cone-rod dystrophy and amelogenesis imperfecta.
Specifications

Specifications

CNNM4
Polyclonal
Unconjugated
CNNM4
Liquid
CNNM4
KLH conjugated synthetic peptide between 565-593 amino acids from the central region of human CNNM4
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.09% sodium azide
Q6P4Q7
ACDP4, ancient conserved domain protein 4, ancient conserved domain-containing protein 4, cyclin-M4, metal transporter CNNM4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26504
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit