Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT4 Rabbit anti-Bovine, Chicken, Human, Mouse, Rat, Xenopus laevis, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523243

Catalog No. PIPA523243


Add to cart

Description

Description

CNOT4 is a 575 amino acid protein with E3 ubiquitin ligase activity, mainly involved in protein modification and protein ubiquitination. It contains a ring type zinc finger domain of the C4C4 type, an RNA recognition motif and a bipartite nuclear localization signal. Studies show that CNOT4 is a component of the CCR4-NOT (CAF1) core complex that functions as general transcription regulation complex and has both a positive as well as a negative effect on the transcription of multiple functionally unrelated genes, thus controls gene expression. CNOT4 interacts with CNOT1 via C-terminus and with E2 ubiquitin ligases via C4C4 RING domain. Reports suggest that CNOT4 gets phosphorylated in response to DNA damage on consensus sites recognized by ATM and ATR.
Specifications

Specifications

CNOT4
Polyclonal
Unconjugated
CNOT4
Liquid
CNOT4, cnot4.L
Synthetic peptide (68 KPLSQEELQRIKNEKKQK 85) of CNOT4 protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Bovine, Chicken, Human, Mouse, Rat, Xenopus laevis
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 0.05% BSA and 0.05% sodium azide
O95628, Q8BT14
CLONE243, NOT4, NOT4H, CCR4-NOT transcription complex subunit 4, CCR4-associated factor 4, E3 ubiquitin-protein ligase CNOT4, NOT4 (negative regulator of transcription 4, yeast) homolog, potential transcriptional repressor NOT4Hp
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
312227, 380304, 417936, 4850, 53621, 540891
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit