Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNOT4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA511532

Catalog No. PIPA511532

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

CNOT4 has intrinsic E3 ubiquitin ligase activity. Binding of the CNOT4 RING finger to the ubiquitin-conjugating enzyme (E2) UbcH5B is highly selective.The CCR4-NOT complex functions as a global regulator of RNA polymerase II transcription.
Specifications

Specifications

CNOT4
Polyclonal
PBS with 0.09% sodium azide
O95628
E3 ubiquitin-protein ligase CNOT4; CCR4-associated factor 4; Potential transcriptional repressor NOT4Hp
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4850
Western Blot
Unconjugated
CNOT4
Liquid
CNOT4
KLH conjugated synthetic peptide between 378-407 amino acids from the C-terminal region of human CNOT4
Ammonium sulfate precipitation, Size-exclusion - Dialysis
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit