Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPase Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA527972

Catalog No. PIPA527972


Add to cart

Description

Description

PA5-27972 targets CNP in WB applications and shows reactivity with Human and mouse samples. The PA5-27972 immunogen is recombit fragment corresponding to a region within amino acids 122 and 382 of Human CNP. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

CNPase
Polyclonal
Unconjugated
CNP
Liquid
CNP
Recombit fragment corresponding to a region within amino acids 122 and 382 of Human CNP
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
P09543, P16330
CNP1, 2′, 3′ cyclic nucleotide 3′ phosphohydrolase, 2′,3′-cyclic-nucleotide 3′-phosphodiesterase, CNPase
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1267, 12799
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit