anti-CNPase, Clone: 11-5B, Millipore™ (Chemicon™)

Manufacturer: EMD Millipore MAB326R/DEL

 View more versions of this product

Catalog No. 50-175-225

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. 

  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Antigen CNPase
    Format Semi-purified
    Host Species Mouse
    Immunogen Purified human 2', 3'-cyclic nucleotide 3'-phosphodiesterase
    Isotype IgG1
    Quantity 100μg
    Regulatory Status RUO
    Research Discipline Neuroscience
    Primary or Secondary Primary
    Monoclonal or Polyclonal Monoclonal
    Conjugate Unlabeled
    Cross Reactivity Bovine,Canine,Human,Mouse,Porcine,Rabbit,Rat,Sheep

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit