Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CNPase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA146433
Description
This antibody is predicted to react with mouse and rat based on 94% sequence homology. Suggested positive control: antigen standard for CNP (transient overexpression lysate), normal human brain lysate.
CNPase Polyclonal antibody specifically detects CNPase in Human, Mouse samples. It is validated for Western Blot.Specifications
CNPase | |
Polyclonal | |
Unconjugated | |
CNP | |
2', 3' cyclic nucleotide 3' phosphohydrolase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic nucleotide 3' phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; 2',3'-cyclic-nucleotide 3'-phosphodiesterase; CNP; Cnp1; Cnp-1; CNPase; CNPF; CNPI; CNPII; cyclic nucleotide phosphodiesterase 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1267, 12799 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P09543, P16330 | |
CNP | |
Residues 68-84 of human CNPase. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction