Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNPase Rabbit anti-Canine, Human, Mouse, Rabbit, Rat, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA519551

Catalog No. PIPA519551

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA5-19551 targets CNPase in WB applications and shows reactivity with Human, mouse, Rabbit, and Rat samples. The PA5-19551 immunogen is synthetic peptide conjugated to KLH derived from within residues 350 to the C-terminus of Human CNPase. PA5-19551 detects CNPase which has a predicted molecular weight of approximately 46 kDa and 48 kDa.

Specifications

Specifications

CNPase
Polyclonal
Unconjugated
CNP
Liquid
CNP
Synthetic peptide conjugated to KLH derived from within residues 350 to the C-terminus of Human CNPase.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Canine, Human, Mouse, Rabbit, Rat
Immunohistochemistry (Paraffin), Western Blot
0.5 mg/mL
PBS with 1% BSA and 0.02% sodium azide
P09543, P16330, P13233
CNP1, 2′, 3′ cyclic nucleotide 3′ phosphohydrolase; 2′,3′-cyclic-nucleotide 3′-phosphodiesterase; CNPase
Rabbit
IgG
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
103351568, 1267, 12799, 25275, 607694
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit