Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CNTF Mouse anti-Human, Clone: 21809, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA523710

Catalog No. PIMA523710


Add to cart

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS. Endoxin level is <0.10 EU per 1 µg of the antibody by the LAL method.

CNTF (ciliary neurotrophic factor) is a polypeptide hormone with activity that is limited to the nervous system. It plays a role in neurotransmitter synthesis, membrane hyper polarization, and neuronal growth. CNTF promotes development of nervous system, and is involved in muscle morphogenesis. It also plays an important role in minimizing damage to neurons during inflammation. CNTF activates JAK-STAT, and PI3K signaling cascades. The protein is encoded by the gene CNTF located on chromosome 11.
Specifications

Specifications

CNTF
Monoclonal
0.5 mg/mL
PBS with 5% trehalose and No Preservative
P26441
ciliary neurotrophic factor, HCNTF
Mouse
IgG2a
500 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
1270
ELISA, Neutralization, Western Blot
21809
Unconjugated
CNTF
Lyophilized
CNTF
E. coli-derived recombit human CTNF Ala2-Met200
Protein A/G
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit