Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ciliary neurotrophic factor, Mouse, Polyclonal Antibody, Abnova™

Mouse polyclonal antibody raised against a full-length human CNTF protein.

Manufacturer:  Abnova Corporation H00001270B01

Catalog No. 89-005-159


Add to cart

Description

Description

The protein encoded by this gene is a polypeptide hormone whose actions appear to be restricted to the nervous system where it promotes neurotransmitter synthesis and neurite outgrowth in certain neuronal populations. The protein is a potent survival factor for neurons and oligodendrocytes and may be relevant in reducing tissue destruction during inflammatory attacks. A mutation in this gene, which results in aberrant splicing, leads to ciliary neurotrophic factor deficiency, but this phenotype is not causally related to neurologic disease. A read-through transcript variant composed of ZFP91 and CNTF sequence has been identified, but it is thought to be noncoding. Read-through transcription of ZFP91 and CNTF has also been observed in mouse. [provided by RefSeq

Sequence: MAFTEHSPLTPHRRDLCSRSIWLARKIRSDLTALTESYVKHQGLNKNINLDSADGMPVASTDQWSELTEAERLQENLQAYRTFHVLLARLLEDQQVHFTPTEGDFHQAIHTLLLQVAAFAYQIEELMILLEYKIPRNEADGMPINVGDGGLFEKKLWGLKVLQELSQWTVRSIHDLRFISSHQTGIPARGSHYIANNKKM
Specifications

Specifications

ciliary neurotrophic factor
Unconjugated
No additive
NM_000614
HCNTF
Mouse
50 uL
Neurobiology
Y
Polyclonal
Human
Western Blot
Mouse polyclonal antibody raised against a full-length human CNTF protein.
CNTF
Serum
CNTF
CNTF (NP_000605, 1 a.a. ~ 200 a.a) full-length human protein.
RUO
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Primary
1270
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit