Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contactin 1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA526499

Catalog No. PIPA526499


Add to cart

Description

Description

This antibody is predicted to react with bovine, mouse and rat based on sequence homology.

CNTN1 is a member of the immunoglobulin superfamily. It is a glycosylphosphatidylinositol (GPI)-anchored neuronal membrane protein that functions as a cell adhesion molecule. It may play a role in the formation of axon connections in the developing nervous system.
Specifications

Specifications

Contactin 1
Polyclonal
PBS with 0.09% sodium azide
Q12860
F3, GP135, contactin-1, glycoprotein gP135, neural cell surface protein F3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
1272
Western Blot
Unconjugated
CNTN1
Liquid
CNTN1
KLH conjugated synthetic peptide between 635-662 amino acids from the central region of human CNTN1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit