Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

C15orf27 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA524298

Catalog No. PIPA524298

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with non-human primate based on sequence homology.

Specifications

Specifications

C15orf27
Polyclonal
PBS with 0.09% sodium azide
Q2M3C6, Q8BZB3
transmembrane protein C15orf27
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
123591, 244886
Flow Cytometry, Western Blot
Unconjugated
TMEM266
Liquid
TMEM266
KLH conjugated synthetic peptide between 503-531 amino acids from the C-terminal region of human CO027
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit