Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cofilin 2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA522301

Catalog No. PIPA522301


Add to cart

Description

Description

PA5-22301 targets Cofilin 2 in ICC, IF, and WB applications and shows reactivity with Human and mouse samples. The PA5-22301 immunogen is a recombit fragment corresponding to a region within amino acids 1 and 166 of Cofilin 2 (muscle) (Uniprot IDNo.Q9Y281). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Specifications

Specifications

Cofilin 2
Polyclonal
Unconjugated
CFL2
Liquid
CFL2
Recombit fragment corresponding to a region within amino acids 1 and 166 of Cofilin 2 (muscle) (Uniprot IDNo.Q9Y281)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunocytochemistry, Immunofluorescence, Western Blot
0.28 mg/mL
PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Q9Y281, P45591
NEM7, cofilin, muscle isoform, cofilin-2
Rabbit
IgG
100 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
1073, 12632
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit