Learn More
collagen, type I, alpha 1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant COL1A1.
Supplier: Abnova Corporation H00001277A01
Description
This gene encodes the pro-alpha1 chains of type I collagen whose triple helix comprises two alpha1 chains and one alpha2 chain. Type I is a fibril-forming collagen found in most connective tissues and is abundant in bone, cornea, dermis and tendon. Mutations in this gene are associated with osteogenesis imperfecta types I-IV, Ehlers-Danlos syndrome type VIIA, Ehlers-Danlos syndrome Classical type, Caffey Disease and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene. [provided by R. Dalgleish
Sequence: EGSPGRDGSPGAKGDRGETGPAGPPGAPGAPGAPGPVGPAGKSGDRGETGPAGPAGPVGPVGARGPAGPQGPRGDKGETGEQGDRGIKSpecifications
collagen, type I, alpha 1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant COL1A1. | |
COL1A1 | |
OI4 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_000088 | |
COL1A1 | |
COL1A1 (NP_000079.1, 1021 a.a. ∼ 1108 a.a) partial recombinant protein with GST tag. | |
RUO | |
1277 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.