Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Collagen I alpha 1, DyLight 550, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177458R

Catalog No. NBP177458R


Add to cart

Description

Description

Collagen I alpha 1 Polyclonal antibody specifically detects Collagen I alpha 1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Collagen I alpha 1
DyLight 550
50mM Sodium Borate with 0.05% Sodium Azide
Affinity Purified
COL1A1
A synthetic peptide made to the C-terminal portion of the human Collagen I protein (between residues 1300-1350) [UniProt P02452]
139 kDa
0.1 ml
Cytoskeleton Markers, Extracellular Matrix
Primary
1277
Human, Mouse
Immunohistochemistry, Immunohistochemistry (Paraffin)
Immunohistochemistry 1:200, Immunohistochemistry-Paraffin 1:200
P02452
Alpha-1 type I collagen, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, collagen, type I, alpha 1, Collagen1, Collagen-1, OI4, pro-alpha-1 collagen type 1
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C in the dark.
Polyclonal
Mouse. Predicted reactivity in Human.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only