Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Collagen I alpha 1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177457C
Description
Collagen I alpha 1 Polyclonal specifically detects Collagen I alpha 1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
Collagen I alpha 1 | |
Polyclonal | |
Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 | |
P02452 | |
COL1A1 | |
This Collagen I alpha 1 antibody was raised against a synthetic peptide made to an internal portion of the human Collagen I protein (between residues 150-200) [UniProt# P02452] | |
Affinity Purified | |
RUO | |
Primary | |
Mouse. Predicted reactivity in Human. | |
Store at 4C in the dark. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Alpha-1 type I collagen, COL-IA1, Collagen 1, Collagen 1 alpha 1, collagen alpha 1 chain type I, collagen alpha-1(I) chain, collagen of skin, tendon and bone, alpha-1 chain, Collagen Type I Alpha 1 Chain, collagen, type I, alpha 1, Collagen1, Collagen-1, EDSARTH1, OI4, pro-alpha-1 collagen type 1, Type I Procollagen Alpha 1 Chain | |
Rabbit | |
139 kDa | |
0.1 mL | |
Cell Biology, Cellular Markers, Extracellular Matrix, Signal Transduction, Stem Cells | |
1277 | |
Human, Mouse | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction