Learn More
Collagen Pan Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA136058
Catalog No. 01-672-241
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 1 mL of distilled water and add preservative if preferred. Antibody has been shown to work well in western blotting under non-reducing conditions.
Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
Specifications
Specifications
Collagen Pan
Polyclonal
whole serum with dextran, mannitol and no preservative
P02452, P02458, P02461, P02462, P05997, P08123, P08572, P20908, P25940, P29400, P53420, Q14031
COL1A1, COL1A2, COL2A1, COL3A1, COL4A1, COL4A2, COL4A4, COL4A5, COL4A6, COL5A1, COL5A2, COL5A3
Purified native collagen type I, II, III, IV and V.
Ammonium sulfate precipitation, Ion-exchange chromatography
RUO
Antibody
Polyclonal
Human
ELISA, Immunohistochemistry (Frozen), Western Blot
Unconjugated
COL2A1
Lyophilized
Rabbit
IgG
1 mL
4° C
Primary
1277, 1278, 1280, 1281, 1282, 1284, 1286, 1287, 1288, 1289, 1290, 50509