Learn More
Collagen I/III Rabbit anti-Bovine, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA135035
Catalog No. 01672493
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Reconstitute with 1 mL of distilled water and add preservative if preferred.
Type I collagen, the fibrillar collagen found in most connective tissues, and the only component of the collagen found in cartilage. Mutations in this gene are associated with osteogenesis imperfecta, Ehlers-Danlos syndrome, and idiopathic osteoporosis. Reciprocal translocations between chromosomes 17 and 22, where this gene and the gene for platelet-derived growth factor beta are located, are associated with a particular type of skin tumor called dermatofibrosarcoma protuberans, resulting from unregulated expression of the growth factor. Two transcripts, resulting from the use of alternate polyadenylation signals, have been identified for this gene.
Specifications
Specifications
Collagen I/III
Polyclonal
Antibody with no preservative
P02453, P02465, P04258
COL1A1, COL1A2, COL3A1, Collagen 1, Collagen 3
Rabbit
IgG
100 μg
4° C
Primary
282187, 282188, 510833
ELISA, Immunohistochemistry (Frozen)
Unconjugated
COL1A2
Lyophilized
COL1A1, COL1A2, COL3A1
Purified bovine native collagen type I and III.
Affinity chromatography
RUO
Antibody
Polyclonal
Bovine